Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509872.2 | XP_003509920.2 | 3 - 4 | 1586 | 107 | |
XM_007650523.1 | XP_007648713.1 | 3 - 4 | 1586 | 107 | ||
XM_007650524.1 | XP_007648714.1 | 3 - 4 | 1569 | 107 | ||
XM_007650525.1 | XP_007648715.1 | 2 - 3 | 1470 | 107 | ||
XM_007650526.1 | XP_007648716.1 | 3 - 4 | 1512 | 107 | ||
GenBank mRNA | JI882005.1 | - | 1 - 2 | 45 | 107 | |
JP062323.1 | - | 1 - 2 | 45 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |