Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652284.1 | XP_007650474.1 | 4 - 5 | 1220 | 98 | |
XM_007652285.1 | XP_007650475.1 | 5 - 6 | 674 | 98 | ||
XM_007652286.1 | XP_007650476.1 | 4 - 5 | 583 | 98 | ||
XM_007652287.1 | XP_007650477.1 | 4 - 5 | 612 | 98 | ||
XM_007652288.1 | XP_007650478.1 | 4 - 5 | 1132 | 98 | ||
GenBank mRNA | JI878795.1 | - | 2 - 3 | 224 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |