Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650953.1 | XP_007649143.1 | 9 - 10 | 2246 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Probe spans exons |