Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643271.1 | XP_007641461.1 | 1 - 2 | 97 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |