Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649890.1 | XP_007648080.1 | 3 - 4 | 384 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |