Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499037.2 | XP_003499085.1 | 1 - 2 | 237 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |