Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509498.2 | XP_003509546.1 | 1 - 2 | 518 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel Probe spans exons |