Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503326.1 | XP_003503374.1 | 3 - 4 | 437 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |