Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ficd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507325.2 | XP_003507373.1 | 2 - 3 | 547 | 121 | |
XM_007647886.1 | XP_007646076.1 | 2 - 3 | 619 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |