Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007615617.1 | XP_007613807.1 | 1 - 2 | 250 | 68 | |
GenBank mRNA | JI880825.1 | - | 1 - 2 | 250 | 68 | |
JP047564.1 | - | 1 - 2 | 250 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |