Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497892.1 | XP_003497940.1 | 8 - 9 | 1009 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |