Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504621.2 | XP_003504669.1 | 2 - 3 | 308 | 84 | |
GenBank mRNA | JI876767.1 | - | 1 - 2 | 50 | 84 | |
JP047664.1 | - | 1 - 2 | 50 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |