Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651496.1 | XP_007649686.1 | 2 - 3 | 1280 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Probe spans exons |