Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499572.2 | XP_003499620.1 | 1 - 2 | 260 | 70 | |
GenBank mRNA | JI868023.1 | - | 1 - 2 | 266 | 70 | |
JP049694.1 | - | 1 - 2 | 266 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |