Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499985.2 | XP_003500033.1 | 2 - 3 | 288 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |