Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509975.2 | XP_003510023.1 | 1 - 2 | 82 | 102 | |
XM_007650640.1 | XP_007648830.1 | 1 - 2 | 82 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |