Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643027.1 | XP_007641217.1 | 7 - 8 | 1492 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Probe spans exons |