Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007626785.1 | XP_007624975.1 | 1 - 2 | 154 | 77 | |
GenBank mRNA | JI870626.1 | - | 1 - 2 | 154 | 77 |