Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633173.1 | XP_007631363.1 | 13 - 14 | 2116 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |