Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504608.1 | XP_003504656.1 | 3 - 4 | 391 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |