Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2l6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509112.2 | XP_003509160.1 | 4 - 5 | 471 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |