Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2l6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509112.2 | XP_003509160.1 | 6 - 7 | 864 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |