Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645767.1 | XP_007643957.1 | 8 - 9 | 1450 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Protein Kinase Probe spans exons |