Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650489.1 | XP_007648679.1 | 1 - 2 | 584 | 75 | |
GenBank mRNA | JI869182.1 | - | 1 - 2 | 503 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |