Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507293.2 | XP_003507341.1 | 1 | 519 | 63 | |
XM_007647856.1 | XP_007646046.1 | 1 | 519 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |