Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513659.2 | XP_003513707.1 | 3 - 4 | 372 | 103 | |
XM_007653918.1 | XP_007652108.1 | 3 - 4 | 302 | 103 | ||
XM_007653919.1 | XP_007652109.1 | 3 - 4 | 469 | 103 | ||
GenBank mRNA | JI891276.1 | - | 3 - 4 | 329 | 103 | |
JP044394.1 | - | 3 - 4 | 329 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |