Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495089.1 | XP_003495137.1 | 5 - 6 | 657 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |