Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506075.2 | XP_003506123.1 | 1 | 1623 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |