Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mei4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510873.2 | XP_003510921.2 | 4 - 5 | 977 | 106 | |
XM_007651459.1 | XP_007649649.1 | 4 - 5 | 966 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |