Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506205.2 | XP_003506253.1 | 2 - 3 | 300 | 80 | |
XM_007646720.1 | XP_007644910.1 | 2 - 3 | 336 | 80 | ||
GenBank mRNA | JI894383.1 | - | 1 - 2 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |