Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514806.2 | XP_003514854.2 | 3 - 4 | 238 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |