Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509681.2 | XP_003509729.1 | 2 - 3 | 176 | 84 | |
GenBank mRNA | JI896129.1 | - | 2 - 3 | 160 | 84 | |
JP056872.1 | - | 2 - 3 | 160 | 84 |