Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640253.1 | XP_007638443.1 | 6 - 7 | 877 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |