Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642471.1 | XP_007640661.1 | 1 - 2 | 679 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |