Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627804.1 | XP_007625994.1 | 5 - 6 | 823 | 91 | |
XM_007627811.1 | XP_007626001.1 | 5 - 6 | 646 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase 3' Most Probe spans exons |