Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651348.1 | XP_007649538.1 | 2 - 3 | 1899 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |