Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511555.2 | XP_003511603.1 | 5 - 6 | 521 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |