Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514154.2 | XP_003514202.1 | 2 - 3 | 645 | 100 | |
XM_007654344.1 | XP_007652534.1 | 2 - 3 | 648 | 100 | ||
XM_007654345.1 | XP_007652535.1 | 2 - 3 | 648 | 100 | ||
XM_007654346.1 | XP_007652536.1 | 2 - 3 | 649 | 100 | ||
XM_007654347.1 | XP_007652537.1 | 2 - 3 | 534 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |