Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653022.1 | XP_007651212.1 | 2 | 671 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |