Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500768.2 | XP_003500816.1 | 2 - 3 | 151 | 80 | |
GenBank mRNA | JI866480.1 | - | 1 - 2 | 87 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |