Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007608675.1 | XP_007606865.1 | 1 - 2 | 85 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Inventoried Probe spans exons |