Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007612568.1 | XP_007610758.1 | 2 - 3 | 515 | 116 | |
GenBank mRNA | JP047629.1 | - | 2 - 3 | 107 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |