Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498019.2 | XP_003498067.1 | 3 - 4 | 648 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |