Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100755558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502912.2 | XP_003502960.1 | 2 - 3 | 426 | 72 | |
XM_007643529.1 | XP_007641719.1 | 2 - 3 | 426 | 72 | ||
GenBank mRNA | JI893402.1 | - | 2 - 3 | 434 | 72 | |
JP043948.1 | - | 2 - 3 | 434 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |