Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503397.1 | XP_003503445.1 | 4 - 5 | 478 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |