Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514821.2 | XP_003514869.1 | 4 - 5 | 922 | 78 | |
XM_007654875.1 | XP_007653065.1 | 4 - 5 | 885 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |