Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514922.2 | XP_003514970.1 | 3 - 4 | 400 | 118 | |
XM_007654957.1 | XP_007653147.1 | 3 - 4 | 403 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |