Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500680.2 | XP_003500728.1 | 2 - 3 | 828 | 73 | |
GenBank mRNA | JI878683.1 | - | 2 - 3 | 395 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |