Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646649.1 | XP_007644839.1 | 8 - 9 | 4896 | 57 | |
XM_007646650.1 | XP_007644840.1 | 8 - 9 | 5298 | 57 | ||
GenBank mRNA | JI879023.1 | - | 1 - 2 | 159 | 57 | |
JP064318.1 | - | 1 - 2 | 155 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |