Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apopt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498760.2 | XP_003498808.2 | 4 - 5 | 401 | 84 | |
GenBank mRNA | JP064365.1 | - | 1 - 2 | 64 | 84 |